missing translation for 'onlineSavingsMsg'
Learn More

GNF179, MedChemExpress

Product Code. 16489487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489487 5 mg 5mg
16426948 10 mg 10mg
16417026 100 mg 100mg
16403007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489487 Supplier Medchem Express Supplier No. HY15975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNF179 is an optimized 8,8-dimethyl IP analog that exhibited the potency(4.8 nM against the multidrug resistant strain W2) in vitro metabolic stability and in vivo oral bioavailability.

GNF179 is an optimized 8,8-dimethyl IP analog that exhibited the potency(4.8 nM against the multidrug resistant strain W2) in vitro metabolic stability and in vivo oral bioavailability.

Specifications

Chemical Name or Material GNF179
CAS 1261114-01-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H23ClFN5O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (233.70 mM; Need ultrasonic)
SMILES CC1(C)C2=NC(C3=CC=C(F)C=C3)=C(NC4=CC=C(Cl)C=C4)N2CCN1C(CN)=O
Molecular Weight (g/mol) 427.9
Formula Weight 427.9
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.