missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GNF 7
GREENER_CHOICE

Product Code. 18743979
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18743979

Brand: Tocris Bioscience™ 5607/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ras signaling inhibitor; inhibits Ack1 and GCK

GNF 7 is a potent Bcr-Abl inhibitor (IC50 values are 61 and 133 nM for T315I and wild-type Bcr-Abl, respectively), inhibiting a range of clinically relevant Bcr-Abl mutants such as T315I, G250E, E255V, F317L and M351T. GNF 7 is also a Ras signaling inhibitor as well as inhibiting Ack1 and germinal center kinase (GCK). GNF 7 induces cell cycle arrest and apoptosis in leukemia cells harboring NRAS mutations, and shows growth inhibitory activity in human colon cancer cells. It prolongs survival in a leukemia cell xenotransplantation model in mice and is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 839706-07-9
Quantity 10 mg
Target Small G Protein Inhibitors
Molecular Formula C28H24F3N7O2
Formula Weight 547.53
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.