missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GNF 5837
GREENER_CHOICE

Product Code. 30001587
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001587 10 mg 10mg
30001612 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001587 Supplier Tocris Bioscience™ Supplier No. 4559/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Trk inhibitor; inhibits TrkA, TrkB and TrkC

Potent pan-Trk inhibitor. Displays antiproliferative effects in cellular Ba/F3 assays (IC50 values are 7, 9 and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively). Exhibits selectivity for Trk receptors over a range of kinases, with some activity at PDGFR and c-Kit (IC50 values are 0.87 and 0.91 μM respectively). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1033769-28-6
Molecular Formula C28H21F4N5O2

Specifications

Quantity 10 mg
Formula Weight 535.49
Purity >98%
Molecular Weight (g/mol) 535.50
Chemical Name or Material GNF 5837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.