missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNF-5837, MedChemExpress
Description
GNF-5837 is a potent, selective, and orally bioavailable pan-tropomyosin receptor kinase (TRK) inhibitor which display antiproliferative effects in cellular Ba/F3 assays ( IC50 values of 7 nM, 9 nM and 11 nM for cells containing the fusion proteins Tel-TrkC, Tel-TrkB and Tel-TrkA, respectively) .
Specifications
Specifications
| Chemical Name or Material | GNF-5837 |
| CAS | 1033769-28-6 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C28H21F4N5O2 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 32 mg/mL (59.76 mM) |
| SMILES | O=C(NC1=CC(C(F)(F)F)=CC=C1F)NC2=CC=C(C)C(NC3=CC(NC/4=O)=C(C=C3)C4=C\C5=CC=CN5)=C2 |
| Molecular Weight (g/mol) | 535.49 |
| Formula Weight | 535.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?