missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GNE 9278
GREENER_CHOICE

Product Code. p-7166778
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18739636

Brand: Tocris Bioscience™ 6369/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Positive allosteric modulator of NMDA receptors; acts in transmembrane domain

GNE 9278 is a positive allosteric modulator of NMDA receptors. Potentiates NMDA receptors containing GluN2A, 2B, 2C and 2D subunits (EC50 values are 0.74, 3.07, 0.47 and 0.32 μM in a Ca2+ influx assay, respectively). Binds within the transmembrane domain and displays preference for agonist-bound NMDA receptors. Slows receptor deactivation and enhances the potency of glutamate and glycine .
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C21H27N5O3S
Formula Weight 429.54
Product Type Positive Allosteric Modulator
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.