missing translation for 'onlineSavingsMsg'
Learn More

GNE-781, MedChemExpress

Product Code. 16475166
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475166 100 mg 100mg
16416947 5 mg 5mg
16404228 10 mg 10mg
16400567 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475166 Supplier Medchem Express Supplier No. HY108696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-781 is an orally active, highly potent and selective CBP inhibitor with an IC50 of 0.94 nM in TR-FRET assay. GNE-781 also inhibits BRET and BRD4(1) with IC50s of 6.2 nM and 5100 nM, respectively. GNE-781 displays antitumor activity in an MOLM-16 AML xenograft model.

GNE-781 is an orally active, highly potent and selective CBP inhibitor with an IC50 of 0.94 nM in TR-FRET assay. GNE-781 also inhibits BRET and BRD4(1) with IC50s of 6.2 nM and 5100 nM, respectively. GNE-781 displays antitumor activity in an MOLM-16 AML xenograft model.

Specifications

Chemical Name or Material GNE-781
Physical Form Solid
CAS 1936422-33-1
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H33F2N7O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (190.26 mM; Need ultrasonic)
SMILES O=C(N1CCC(N(C2CCOCC2)N=C3N4CCCC5=C4C=C(C(F)F)C(C6=CN(C)N=C6)=C5)=C3C1)NC
Molecular Weight (g/mol) 525.59
Formula Weight 525.59
Percent Purity 98.19%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.