missing translation for 'onlineSavingsMsg'
Learn More

GNE-477, MedChemExpress

Product Code. 16494378
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494378 10 mg 10mg
16483056 25 mg 25mg
16441149 1 mg 1mg
16437107 5 mg 5mg
16410707 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494378 Supplier Medchem Express Supplier No. HY11042
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.

GNE-477 is a potent and efficacious dual PI3K (IC50=4 nM)/mTOR(Ki=21 nM) inhibitor.

Specifications

Chemical Name or Material GNE-477
CAS 1032754-81-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H28N8O3S2
Quantity 10 mg
Solubility Information DMSO : 16.67 mg/mL (33.03 mM; Need ultrasonic)
SMILES NC1=NC=C(C2=NC(N3CCOCC3)=C4C(C(C)=C(CN5CCN(S(=O)(C)=O)CC5)S4)=N2)C=N1
Molecular Weight (g/mol) 504.63
Formula Weight 504.63
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.