missing translation for 'onlineSavingsMsg'
Learn More

GNE-272, MedChemExpress

Product Code. 16475867
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475867

Brand: Medchem Express HY100726

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.

GNE-272 is a potent and selective CBP/EP300 inhibitor with IC50 values of 0.02, 0.03 and 13 μM for CBP, EP300 and BRD4, respectively. GNE-272 is also a selective in vivo probe for CBP/EP300.

Specifications

Chemical Name or Material GNE-272
CAS 1936428-93-1
Color White
Purity Grade Notes Research
Molecular Formula C22H25FN6O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (235.59 mM; Need ultrasonic)
SMILES CC(N1CCC2=C(C(NC3=C(F)C=C(C4=CN(C)N=C4)C=C3)=NN2[C@@H]5COCC5)C1)=O
Molecular Weight (g/mol) 424.47
Formula Weight 424.47
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.