missing translation for 'onlineSavingsMsg'
Learn More

GNE-131, MedChemExpress

Product Code. 16494608
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494608 10 mg 10mg
16490907 50 mg 50mg
16465456 100 mg 100mg
16447327 5 mg 5mg
16423166 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494608 Supplier Medchem Express Supplier No. HY112279
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GNE-131 is a potent and selective inhibitor of human sodium channel NaV1.7, with an IC50 of 3 nM.

GNE-131 is a potent and selective inhibitor of human sodium channel NaV1.7, with an IC50 of 3 nM.

Specifications

Chemical Name or Material GNE-131
CAS 1629063-81-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H30N4O3S
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (282.44 mM; Need ultrasonic)
SMILES O=S(NC1=NN=C2N1C=C(C3CC3)C(OCC45C[C@@H](C6)C[C@@H](C[C@@H]6C5)C4)=C2)(C7CC7)=O
Molecular Weight (g/mol) 442.57
Formula Weight 442.57
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.