missing translation for 'onlineSavingsMsg'
Learn More

GNE-049, MedChemExpress

Product Code. 16445156
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16445156 100 mg 100mg
16434198 10 mg 10mg
16416917 5 mg 5mg
16410547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16445156 Supplier Medchem Express Supplier No. HY108435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GNE-049 is a highly potent and selective CBP inhibitor with an IC50 of 1.1 nM in TR-FRET assay. GNE-049 also inhibits BRET and BRD4(1) with IC50s of 12 nM and 4200 nM, respectively.

GNE-049 is a highly potent and selective CBP inhibitor with an IC50 of 1.1 nM in TR-FRET assay. GNE-049 also inhibits BRET and BRD4(1) with IC50s of 12 nM and 4200 nM, respectively.

Specifications

Chemical Name or Material GNE-049
Physical Form Solid
CAS 1936421-41-8
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H32F2N6O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (195.86 mM; Need ultrasonic)
SMILES CC(N1CCC(N(C2CCOCC2)N=C3N4CCCC5=C4C=C(C(F)F)C(C6=CN(C)N=C6)=C5)=C3C1)=O
Molecular Weight (g/mol) 510.58
Formula Weight 510.58
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.