missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNB5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101627
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GNB5.
The Gb5 protein is a G protein subunit present predominantly in the central nervous system. This protein appears to be expressed in two forms, one found in the brain of 39 kDa and one found in the retina of 44 kDa. It has also been found that Gb5 associates tightly with RGS7, a member of the superfamily of negative regulators of G protein signaling. These complexes have been localized to most regions of the brain, whereas Gb1-4 exhibit no observable colocalization with RGS7, despite their widespread codistribution throughout similar neuronal networks. This antibody is specific for a peptide sequence within the D1 blade which is not found in the Gb's 1-4.
Specifications
| GNB5 | |
| Polyclonal | |
| Unconjugated | |
| Gnb5 | |
| flail; flailer; flr; G protein subunit beta 5; G protein, beta subunit 5L; G protein, beta-5 subunit; GB5; GBB5; Gbeta5; GBS; Gnb5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; transducin beta chain 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10681, 14697, 83579 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14775, P62881, P62882 | |
| Gnb5 | |
| A synthesized peptide derived from human GNB5(Accession O14775), corresponding to amino acid residues V240-Y290. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction