missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GNB5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30850-100ug
This item is not returnable.
View return policy
Description
GNB5 Polyclonal antibody specifically detects GNB5 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| GNB5 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| FLJ37457, FLJ43714, G protein, beta subunit 5L, G protein, beta-5 subunit, GB5, gbeta5, guanine nucleotide binding protein (G protein), beta 5, guanine nucleotide-binding protein subunit beta-5, guanine nucleotide-binding protein, beta subunit 5L, Transducin beta chain 5 | |
| Rabbits were immunized with a unique peptide sequence derived from the D1 blade of the G 5 protein | |
| 100 μg | |
| Signal Transduction | |
| 10681 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction