missing translation for 'onlineSavingsMsg'
Learn More

GNB5 Antibody, HRP, Novus Biologicals™

Product Code. 18399451 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399451 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399451 Supplier Novus Biologicals Supplier No. NBP306495H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GNB5 Polyclonal antibody specifically detects GNB5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNB5
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias FLJ37457, FLJ43714, G protein, beta subunit 5L, G protein, beta-5 subunit, GB5, gbeta5, guanine nucleotide binding protein (G protein), beta 5, guanine nucleotide-binding protein subunit beta-5, guanine nucleotide-binding protein, beta subunit 5L, Transducin beta chain 5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GNB5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10681
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.