missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16090-1-AP-150UL
This item is not returnable.
View return policy
Description
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene contains a trinucleotide (CCG) repeat length polymorphism in its 5′ UTR.Specifications
| GNB2 | |
| Polyclonal | |
| Unconjugated | |
| GNB2 | |
| G protein subunit beta 2, GNB2, Transducin beta chain 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14693, 2783, 81667 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62879, P62880 | |
| GNB2 | |
| GNB2 Fusion Protein Ag9125 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion