missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16090-1-AP-20UL
This item is not returnable.
View return policy
Description
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene contains a trinucleotide (CCG) repeat length polymorphism in its 5′ UTR.Specifications
| GNB2 | |
| Polyclonal | |
| Unconjugated | |
| GNB2 | |
| G protein subunit beta 2, GNB2, Transducin beta chain 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14693, 2783, 81667 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62879, P62880 | |
| GNB2 | |
| GNB2 Fusion Protein Ag9125 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction