Learn More
GNB1L Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26484-1-AP-150UL
This item is not returnable.
View return policy
Description
The GNB1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GNB1L. This antibody recognizes human antigen. The GNB1L antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GNB1L | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| DGCRK3, FKSG1, GNB1L, GY2, KIAA1645, WDR14, WDVCF | |
| 150 μL | |
| 54584 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |