missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58342
This item is not returnable.
View return policy
Description
GNB1L Polyclonal specifically detects GNB1L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GNB1L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DGCRK3, G-protein beta subunit-like protein, guanine nucleotide binding protein (G protein), beta polypeptide 1-like, guanine nucleotide binding protein beta-subunit-like polypeptide, guanine nucleotide-binding protein subunit beta-like protein 1, GY2WD40 repeat-containing protein deleted in VCFS, KIAA1645, WD repeat-containing protein 14, WDR14G protein subunit beta-like protein 1, WDVCF | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: beta-like protein 1. | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9BYB4 | |
| GNB1L | |
| Synthetic peptides corresponding to GNB1L (guanine nucleotide binding protein (G protein), beta polypeptide 1-like) The peptide sequence was selected from the C terminal of GNB1L. Peptide sequence RVFHWRTMQPLAVLAFHSAAVQCVAFTADGLLAAGSKDQRISLWSLYPRA The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Signal Transduction | |
| 54584 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction