missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNAT1 Recombinant Rabbit Monoclonal Antibody (HL2126)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556725
This item is not returnable.
View return policy
Description
GNAT1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phosphodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in rods. This gene is also expressed in other cells, and has been implicated in bitter taste transduction in rat taste cells. Mutations in this gene result in autosomal dominant congenital stationary night blindness. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| GNAT1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P11488, P20612, Q28300 | |
| GNAT1 | |
| Recombinant fragment of human GNAT1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2126 | |
| Unconjugated | |
| GNAT1 | |
| CSNB1G; CSNBAD3; G protein subunit alpha transducin 1; GBT1; Gnat1; Gnat-1; GNATR; guanine nucleotide binding protein (G protein), alpha transducing 1; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; guanine nucleotide binding protein, alpha transducing 1; guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; Ird1; Ird2; irdc; irdr; rod-type transducin alpha subunit; Tr alpha; Tralpha; transducin; transducin alpha subunit; transducin alpha-1 chain; transducin alpha-subunit; transducin, rod-specific; transducin-alpha | |
| Rabbit | |
| Protein A | |
| RUO | |
| 140428, 14685, 2779, 363143, 403613 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction