missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNAI2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143027
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_002061.1; NP_001159897.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
The protein encoded by this gene is an alpha subunit of guanine nucleotide binding proteins (G proteins). The encoded protein contains the guanine nucleotide binding site and is involved in the hormonal regulation of adenylate cyclase. Several transcript variants encoding different isoforms have been detected for this gene, but the full-length nature of only two are known so far.
Specifications
| GNAI2 | |
| Polyclonal | |
| Unconjugated | |
| Gnai2 | |
| adenylate cyclase-inhibiting G alpha protein; BR; C76432; CD49B; G alpha inhibitory type 2 (a); G alpha inhibitory type 2 (b); G protein subunit alpha i2; Galphai2; Gia; GIP; GNAI2; Gnai-2; GNAI2B; GPIa; GTP-binding protein (G-alpha-i2); GTP-binding regulatory protein Gi alpha-2 chain; guanidine nucleotide binding protein, (G protein) , alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein (G protein), alpha inhibiting 2; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2; guanine nucleotide binding protein, alpha inhibiting 2; guanine nucleotide binding protein, alpha inhibiting activity polypeptide 2; guanine nucleotide-binding protein G(i) subunit alpha-2; guanine nucleotide-binding protein G(i), alpha-2 subunit; H_LUCA15.1; H_LUCA16.1; inhibitory GTP-binding protein subunit alpha 2; VLA-2; VLAA2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P04899 | |
| Gnai2 | |
| Peptide with sequence C-PEYTGANKYDE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction