missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNAI1/GNAI2/GNAI3 Recombinant Rabbit Monoclonal Antibody (HL2093)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550379
This item is not returnable.
View return policy
Description
GNAI1/GNAI2/GNAI3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
GNAI1/GNAI2/GNAI3 are genes that encode for the G protein alpha inhibiting activity subunit. These proteins play a crucial role in signal transduction pathways, particularly in the inhibition of adenylyl cyclase. They are involved in various physiological processes, including neurotransmitter release, hormone secretion, and regulation of smooth muscle contraction. Mutations in these genes have been associated with various diseases, including hypoparathyroidism, pituitary adenoma, and cardiovascular disorders.
Specifications
| GNAI1/GNAI2/GNAI3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| B2RSH2, P04897, P04899, P08752, P08753, P08754, P10824, P63096, Q9DC51 | |
| GNAI1, GNAI2, GNAI3 | |
| Recombinant fragment of human GNAI3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2093 | |
| Unconjugated | |
| GNAI3 | |
| HG1B/HG1C/HG1A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100006888, 14677, 14678, 14679, 25643, 25686, 2770, 2771, 2773, 393946, 81664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction