missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNA12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100663
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GNA12.
Guanine nucleotide-binding proteins (G proteins) are modulators or transducers in various transmembrane signaling systems. G proteins are composed of 3 subunits; alpha, beta and gamma. The alpha chain contains the guanine nucleotide binding site.
Specifications
| GNA12 | |
| Polyclonal | |
| Unconjugated | |
| GNA12 | |
| AI414047; AI504261; G alpha-12; G protein subunit alpha 12; Galpha12; gep; GNA12; Gna-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; guanine nucleotide binding protein, alpha 12; guanine nucleotide-binding protein subunit alpha-12; NNX3; RMP; WUGSC:H_GS165O14.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14673, 2768, 81663 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27600, Q03113, Q63210 | |
| GNA12 | |
| A synthesized peptide derived from human GNA12(Accession Q03113), corresponding to amino acid residues N296-L346. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction