missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNA12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597793
This item is not returnable.
View return policy
Description
GNA12 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Guanine nucleotide-binding proteins (G proteins) are modulators or transducers in various transmembrane signaling systems. G proteins are composed of 3 subunits; alpha, beta and gamma. The alpha chain contains the guanine nucleotide binding site.
Specifications
| GNA12 | |
| Polyclonal | |
| Unconjugated | |
| GNA12 | |
| AI414047; AI504261; G alpha-12; G protein subunit alpha 12; Galpha12; gep; GNA12; Gna-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; guanine nucleotide binding protein, alpha 12; guanine nucleotide-binding protein subunit alpha-12; NNX3; RMP; WUGSC:H_GS165O14.2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2768 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.63 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q03113 | |
| GNA12 | |
| Recombinant Human Guanine nucleotide-binding protein subunit alpha-12 protein (112-270AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction