missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) GN-6 Metricel™ Membrane Disc Filters

Certified 0.45 μm membrane for microbiological analysis meets or surpasses regulatory specifications.

Brand:  Cytiva (formerly Pall Lab) 63068

Additional Details : Weight : 0.15200kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 17154531

  • £90.65 / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Mixed cellulose esters is the most accepted filter media for microbiological analysis, and provides maximum recovery of organisms
  • Unique dot grid pattern gridline provides guidance for quantification of bacterial colonies without growth inhibition or enhancement
  • Suitable for microbiological analysis using the Membrane Filter (MF) Technique. Each box contains a signed certification for your permanent records
  • Variety of packaging options meets any need. Available in individually packaged S-Packs, autoclave packs, or non-sterile packs
  • Use with nutrient media for high growth levels and easy identification of contaminants
  • Designed for laboratory applications only
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

Hydrophilic MCE
Non-sterile
152 μm
25 mm
0.45 μm
100/Pk.
Plain
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Membranes: cellulose nitrate and esters of cellulose

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers