missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565452
This item is not returnable.
View return policy
Description
Immunogen sequence: AGGDLKDGHHH YEGAVVILDA GAQYGKVIDR RVRELFVQSE IFPLETPAFA IKEQGFRAII ISGGPNSVYA EDAPWFDPAI FTIGKPVLGI CYGMQMMNKV FGGTVHKKSV REDGVFNISV DNTCSLFRGL QKEEVVLLT Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
Specifications
| GMPS | |
| Polyclonal | |
| Unconjugated | |
| GMPS | |
| AA591640; AI047208; Glutamine amidotransferase; GMP synthase; GMP synthase [glutamine-hydrolyzing]; GMP synthetase; GMPS; guanine monophosphate synthase; guanine monophosphate synthetase; guanine monphosphate synthase; guanine monphosphate synthetase; guanosine 5'-monophosphate synthase; MLL/GMPS fusion protein; testicular tissue protein Li 82 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8833 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49915 | |
| GMPS | |
| Recombinant Human GMPS. Recombinant protein control fragment (Product #RP-106132). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction