missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPR Monoclonal Antibody (3G12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524396
This item is not returnable.
View return policy
Description
Peptide Sequence: MPRIDADLKL DFKDVLLRPK RSSLKSRAEV DLERTFTFRN SKQTYSGIPI IVANMDTVGT FEMAAVMSQH SMFTAIHKHY SLDDWKLFAT NHPECLQNVA VSSGSGQNDL EKMTSILEAV.
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.
Specifications
| GMPR | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with no preservative | |
| P36959 | |
| GMPR | |
| GMPR (NP_006868, 1 a.a. ~ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2766 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3G12 | |
| Unconjugated | |
| GMPR | |
| 2310004P21Rik; AV028449; GMP reductase 1; GMPR; GMPR 1; Gmpr1; guanine monophosphate reductase; guanosine 5'-monophosphate oxidoreductase 1; guanosine monophosphate reductase; guanosine monophosphate reductase 1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction