missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569559
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Goat: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 86%.
This gene is thought to encode a GDP-mannose pyrophosphorylase. The encoded protein catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose, a reaction involved in the production of N-linked oligosaccharides. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| GMPPB | |
| Polyclonal | |
| Unconjugated | |
| Gmppb | |
| 2.7.7.13; AI317178; cb1066; E430010H19; fc79b10; GDP-mannose pyrophosphorylase B; Gmppb; gmppl; GTP-mannose-1-phosphate guanylyltransferase beta; Mannose-1-phosphate guanyltransferase beta; MDDGA14; MDDGB14; MDDGC14; RGD1560458; Unknown (protein for MGC:134517); wu:fc79b10; zgc:92026 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 331026 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8BTZ7 | |
| Gmppb | |
| synthetic peptide directed towards the middle region of human GMPPB (aa 2-51). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction