missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMPPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557303
This item is not returnable.
View return policy
Description
Immunogen sequence: TCVLHSIVGW GSTVGRWARV EGTPSDPNPN DPRARMDSES LFKDGKLLPA ITILGCRVRI PAEVLILNSI VLPHKELSRS FTNQIIL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.
Specifications
| GMPPA | |
| Polyclonal | |
| Unconjugated | |
| GMPPA | |
| 1810012N01Rik; AAMR; GDP-mannose pyrophosphorylase A; Gmppa; GMPP-alpha; GTP-mannose-1-phosphate guanylyltransferase alpha; mannose-1-phosphate guanyltransferase alpha; mannose-1-phosphate guanylyltransferase (GDP) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96IJ6 | |
| GMPPA | |
| Recombinant protein corresponding to Human GMPPA. Recombinant protein control fragment (Product #RP-96134). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction