Learn More
Thermo Scientific™ GMP Startup, Calibration, IQOQ+PT (cycle test), Preventive Maintenance for Plus Centrifuge (New Product)
Product Code. 31703127
Change view
Click to view available options
For Use With (Equipment):
Centrifuges
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31703127 Centrifuges Each
1 options
Product Code. 31703127 Supplier Thermo Scientific™ Supplier No. GMPCENPLUSNP
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This GMP service plan supports centrifuge plus (Superspeed/Ultra) during new product stage. Includes priority CM support (Next business day-select locations), all-inclusive repair, E-learning, no-charge requalification gaurantee, 1 hr. free consultation, dedicated coordinator, ensuring minimized downtime.
GMP Service Plan - New Product (NP) (Part of the Regulatory Assurance Plans)
Overview
  • Tailored for centrifuge plus models (Ultra/Superspeed) at the new equipment stage.
  • Delivers comprehensive startup, qualification, and preventive maintenance support for GMP compliance.
Included Services
  • Start-up: Helps ensure that your new equipment is installed, connected, and fully operational.
  • Calibration: Periodic verification confirms that equipment and monitoring sensors produce accurate results within specified limits. This service maintains compliance with ISO 9001 standards.
  • Installation qualification (IQ): Documented verification that the equipment installation site and environment meet the manufacturer’s specifications and user requirements. IQ verifies that the equipment, manuals, supplies, and any other accessories arrived undamaged and are properly assembled and installed. Configuration details for each system component are recorded.
  • Operational qualification (OQ): Documented verification that the equipment (as installed) is operating as intended according to manufacturer’s specifications. OQ verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer, and that it conforms to standards and requirements. Key tests include door ajar, power failure, and temperature alerts.
  • Performance testing (PT): Custom tests to help ensure that the equipment meets user specifications like cycle testing (CT) for centrifuges.
  • Preventive maintenance: Routine inspections using standard procedures to ensure equipment performance is at or near the same level achieved at installation. Typical procedures include general inspection and cleanup, replacement of worn parts, performance testing, and tuning.
Additional Benefits
  • Priority enhanced support commitment (Next business day) in select locations
  • All-inclusive repair warranty and coverage, including spare parts.
  • Dedicated account coordinator for personalized support.
  • 1 hr. consultation for compliance and service support guidance.
  • Unlimited E-learning access.
  • 50% discount on remote monitoring solutions and optional add-ons (temp map/requalification)
Customer Outcomes
  • Reliable performance and minimized downtime
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Centrifuges
Product Type Service