Learn More
Thermo Scientific™ GMP Startup, Calibration, IQOQ + PT (1 Temp Map if Applied) and Preventive Maintenance for Shaker (New Product)
Product Code. 31703125
Change view
Click to view available options
Product Type:
Service
Unit Size:
Each
Product Code. Product Type Unit Size
31703125 Service Each
1 options
Product Code. 31703125 Supplier Thermo Scientific™ Supplier No. GMPSHKNP
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This GMP service plan supports a shaker during the new product stage. Includes priority CM support (NBD in select locations), all-inclusive parts, repair, E-learning, no-charge requalification guarantee, 1 hr. free consultation, dedicated coordinator, ensuring minimized downtime.
GMP Service Plan - New Product (NP) (Part of the Regulatory Assurance Plans)
Overview
  • Tailored for shakers at the new equipment stage.
  • Delivers comprehensive startup, qualification, and preventive maintenance support for GMP compliance.
Included Services
  • Start-up: Helps ensure that your new equipment is installed, connected, and fully operational.
  • Calibration: Periodic verification confirms that equipment and monitoring sensors produce accurate results within specified limits and maintains compliance with ISO 9001 standards.
  • Installation qualification (IQ): Verification that the equipment installation site and environment meet manufacturer specifications and user requirements. Ensures equipment, manuals, supplies, and accessories arrive undamaged and are properly assembled and installed. Configuration details are recorded.
  • Operational qualification (OQ): Verification that the equipment operates as intended according to manufacturer specifications, including key tests such as door ajar, power failure, and temperature alerts.
  • Performance testing (PT): Custom test to ensure equipment meets user specifications such as 1 temperature mapping.
  • Preventive maintenance: Routine inspections using standard procedures to sustain equipment performance. Includes inspection, cleanup, worn part replacement, performance testing, and tuning.
Additional Benefits
  • Priority enhanced support commitment (Next business day) in select locations
  • All-inclusive repair warranty and coverage, including spare parts.
  • Dedicated account coordinator for personalized support.
  • 1-hour consultation for compliance and service support guidance.
  • Unlimited E-learning access.
  • 50% discount on remote monitoring solutions and optional add-ons (temp map/requalification)
Customer Outcomes
  • Seamless startup with validated documentation.
  • Reliable performance and minimized downtime.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Service