Learn More
R&D Systems™ GMP N-2 MAX Media Supplement (100X), Animal-free
Product Code. 18786506
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. 18786506
Brand: R&D Systems™ AR016
GMP Animal-free Media Supplement for Ex Vivo Cell or Tissue Manufacturing
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 5 mL
Species Multi-Species