missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GMP Human Laminin 511 Protein

Product Code. 31453817
Change view
Click to view available options
Quantity:
10.5 mg
1050 μg
175 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31453817 175 μg
31452279 10.5 mg
31452915 1050 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31453817 Supplier AcroBiosystems Supplier No. GMPLA1H25175ug
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Recombinant Protein

Laminin 511 E8 is a truncated laminin 511, which consists of the C-terminal end of the alpha 5, beta 1, and gamma 1 chains, as one of the ECM proteins. Laminin E8s serve as functionally minimal forms that retain the full capability for binding to integrins. More importantly, laminin 511 E8 is the key matrix for feeder-free culture of human embryonic (hESC) and induced pluripotent stem cells (iPSC). Furthermore, laminin 511 E8 could also support the self-renewal of hESC and iPSC without spontaneous differentiation, in combination with defined stem cell culture media.

Specifications

Format Liquid
Formulation Supplied as 0.2 μm filtered solution in PBS, pH 7.4 with protectants. Contact us for customized product form or formulation.
Quantity 175 μg
Biological Activity GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) immobilized on CM5 Chip can bind Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein, His Tag&Tag Free with an affinity constant between 1.00 nM - 15.0 nM as determined in a SPR assay (Biacore 8K) (QC tested).∣The SPR based assay shows that GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) is stable at 37°C for 23 days.∣The SPR based assay shows that GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) is stable after freezing and thawing 3 times.∣The SPR based assay shows batch-to-batch consistency between Acro's GMP and PG Laminin 511.∣GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) and Human Laminin 511 Protein, premium grade (Cat. No. LA8-H5283L) could maintain the stemness of iPSC at least Passage 5 and has similar performance. FACS data indicated that the iPSCs expressed high levels of pluripotency associated markers OCT4, SOX2, and SSEA4.∣GMP Human Laminin 511 Protein (Cat. No. GMP-LA1H25) and Human Laminin 511 Protein, premium grade (Cat. No. LA8-H5283L) could maintain the stemness of iPSC at least Passage 5 and have similar performance. Immunofluorescence staining indicated that the iPSCs expressed high levels of pluripotency associated markers OCT4, SOX2, and NANOG.
Species Human
Content And Storage 2°C to 8°C for 6 months under sterile conditions; -20°C or below for 3 years. For long term storage, the product should be stored at liquid state at -20°C or below.
Shipping Condition This product is supplied and shipped with dry ice, please inquire the shipping cost.
Purity or Quality Grade 0.95
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.