missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMNN Recombinant Rabbit Monoclonal Antibody (24GB4610)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | GMNN |
|---|---|
| Clone | 24GB4610 |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276036
![]() |
Invitrogen™
MA557029 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276492
|
Invitrogen™
MA557030 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GMNN Recombinant Monoclonal Antibody for Western Blot
GMNN inhibits DNA replication by preventing the incorporation of MCM complex into prereplication complex (pre-RC). It is degraded during the mitotic phase of the cell cycle. Its destruction at the metaphase-anaphase transition permits replication in the succeeding cell cycle.Specifications
| GMNN | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| GMNN | |
| AW546347; Gem; geminin; Geminin GMNN; geminin, DNA replication inhibitor; GMNN; MGORS6; OTTHUMP00000039393; RP3 369A17.3 | |
| GMNN | |
| Primary | |
| Affinity chromatography |
| 24GB4610 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75496 | |
| 291137, 51053 | |
| Recombinant protein of human Geminin (aa 110-209). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title