missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GML Antibody (1E7), Novus Biologicals™

Product Code. 18346398 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346398 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346398 Supplier Novus Biologicals™ Supplier No. H00002765M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GML Monoclonal antibody specifically detects GML in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GML
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002057
Gene Alias glycosylphosphatidylinositol anchored molecule like protein, glycosyl-phosphatidylinositol-anchored molecule-like protein, Glycosylphosphatidylinositol-anchored molecule-like protein, LY6DLGPI anchored molecule like protein
Host Species Mouse
Immunogen GML (NP_002057, 48 a.a. ∽ 158 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CPYHIRRCMTISIRINSRELLVYKNCTNNCTFVYAAEQPPEAPGKIFKTNSFYWVCCCNSMVCNAGGPTNLERDMLPDEVTEEELPEGTVRLGVSKLLLSFASIIVSNILP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 2765
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.