missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMFG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13625-1-AP-20UL
This item is not returnable.
View return policy
Description
Glia maturation factor gamma (GMFG) is a cytokine-responsive protein in erythropoietin-induced and granulocyte-colony stimulating factor-induced hematopoietic lineage development. It may be involved in glial differentiation, neural regeneration, and inhibition of tumor cell proliferation.Specifications
| GMFG | |
| Polyclonal | |
| Unconjugated | |
| GMFG | |
| Glia maturation factor gamma, glia maturation factor, gamma, GMF GAMMA, GMFG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113940, 63986, 9535 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60234, Q80T18, Q9ERL7 | |
| Gmfg | |
| GMFG Fusion Protein Ag4538 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction