missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMFG Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550345
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total GMFG protein.
Glia maturation factor gamma (GMFG) is a cytokine-responsive protein in erythropoietin-induced and granulocyte-colony stimulating factor-induced hematopoietic lineage development. It may be involved in glial differentiation, neural regeneration, and inhibition of tumor cell proliferation.
Specifications
| GMFG | |
| Polyclonal | |
| Unconjugated | |
| Gmfg | |
| 0610039G16Rik; 2310057N07Rik; AI324845; glia maturation factor gamma; glia maturation factor, gamma; Gmfg; GMF-gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63986, 9535 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O60234, Q9ERL7 | |
| Gmfg | |
| Full length fusion protein of Human GMFG | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction