missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMF-beta Antibody (44D8), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59490G
This item is not returnable.
View return policy
Description
GMF-beta Monoclonal antibody specifically detects GMF-beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| GMF-beta | |
| Monoclonal | |
| DyLight 488 | |
| glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
| Recombinant human GMF-beta (1-142aa) purified from E. coli | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 2764 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 44D8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction