missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMEB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13314-1-AP-150UL
This item is not returnable.
View return policy
Description
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. While little is known about ZBTB40, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Single nucleotide polymorphisms near ZBTB40 have been linked to differences in bone mineral density of the hip and lumbar spine, and had a significant association with low-trauma fractures suggesting that ZBTB40 may be involved in bone development and formation.Specifications
| GMEB2 | |
| Polyclonal | |
| Unconjugated | |
| GMEB2 | |
| DNA binding protein p79PIF, GMEB 2, GMEB2, KIAA1269, P79PIF, PIF p79, PIF79 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229004, 26205, 83635 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88873, P58929, Q9UKD1 | |
| Gmeb2 | |
| GMEB2 Fusion Protein Ag4157 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction