missing translation for 'onlineSavingsMsg'
Learn More

GMEB2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18416385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416385 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416385 Supplier Novus Biologicals Supplier No. NBP212763AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GMEB2 Polyclonal antibody specifically detects GMEB2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GMEB2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DNA-binding protein p79PIF, glucocorticoid modulatory element binding protein 2, KIAA1269PIF p79, p79, P79PIF
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 480 and 530 of human Glucocorticoid Modulatory Element Binding Protein 2 using the numbering given in entry NP_036516.1 (GeneID 26205).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26205
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.