missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GMEB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584134
This item is not returnable.
View return policy
Description
Immunogen sequence: STLGNMTTMV SPVELVAMES GLTSAIQAVE STSEDGQTII EIDPAPDPEA EDTEGKAVIL ETELRTEEKV VAEMEEHQHQ VHNVEI Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.
This gene is a member of KDWK gene family. The product of this gene associates with GMEB2 protein, and the complex is essential for parvovirus DNA replication. Study of rat homolog implicates the role of this gene in modulation of transactivation by the glucocorticoid receptor bound to glucocorticoid response elements. Two alternative spliced transcript variants encoding different isoforms exist.
Specifications
| GMEB1 | |
| Polyclonal | |
| Unconjugated | |
| GMEB1 | |
| 1110050A04Rik; AI256615; AI481278; DNA-binding protein p96PIF; glucocorticoid modulatory element binding protein 1; glucocorticoid modulatory element-binding protein 1; Gmeb1; GMEB-1; P96PIF; Parvovirus initiation factor p96; PIF p96; PIF96; RGD1563208 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y692 | |
| GMEB1 | |
| Recombinant protein corresponding to Human GMEB1. Recombinant protein control fragment (Product #RP-106431). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction