missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMDS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568194
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Catalyzes the conversion of GDP-D-mannose to GDP-4-dehydro-6-deoxy-D-mannose.
Specifications
| GMDS | |
| Polyclonal | |
| Unconjugated | |
| GMDS | |
| RP1-118B18.1, GMD, SDR3E1, GDP-D-mannose dehydratase, GDP-mannose 4,6 dehydratase, short chain dehydrogenase/reductase family 3E, member 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 218138, 2762, 291095 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60547, Q8K0C9 | |
| GMDS | |
| Synthesized peptide derived from human GMDS. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction