missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMDS Antibody (OTI2A1), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70835AF647
This item is not returnable.
View return policy
Description
GMDS Monoclonal specifically detects GMDS in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GMDS | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GMDS | |
| Full length recombinant protein of human GMDS (NP_001491) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI2A1 | |
| Western Blot | |
| EC 4.2.1.47, GDP-D-mannose dehydratase, GDP-mannose 4,6-dehydratase, GMDGDP-mannose 4,6 dehydratase, SDR3E1, short chain dehydrogenase/reductase family 3E, member 1 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 2762.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction