missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ GMB 475
Description
GMB 475 is a BCR-ABL1 kinase PROTAC™ Degrader. Induces degradation of BCR-ABL1 and cABL1, inhibits downstream STAT5 signaling and suppresses cell proliferation in chronic myeloid leukemia cells lines. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-3,3-Dimethyl-2-(2-(2-(4-(6-((4-(trifluoromethoxy)phenyl)amino)pyrimidin-4-yl)phenoxy)ethoxy)acetamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2490599-18-1 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Active Degraders |
| Molecular Formula | C43H46F3N7O7S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?