missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GM632 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-28902
This item is not returnable.
View return policy
Description
GM632 Polyclonal antibody specifically detects GM632 in Human samples. It is validated for Western Blot
Specifications
| GM632 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57473 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GM632, KIAA1196, MGC149845, MGC149846, zinc finger protein 512B | |
| between 650 and 700 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction