missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2A Mouse anti-Human, Clone: 1E4D3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GM2A |
|---|---|
| Clone | 1E4D3 |
| Concentration | 1.64 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817635
|
Proteintech
66080-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807635
|
Proteintech
66080-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta-hexosaminidase A. Beta-hexosaminidase A, together with GM2 ganglioside activator, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Mutations in this gene result in GM2-gangliosidosis type AB or the AB variant of Tay-Sachs disease. Alternative splicing results in multiple transcript variants.Specifications
| GM2A | |
| 1.64 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Ganglioside GM2 activator, GM2 AP, GM2 ganglioside activator, GM2A, SAP 3 | |
| GM2A | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1E4D3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P17900 | |
| 2760 | |
| GM2A Fusion Protein Ag4394 | |
| Primary | |
| -20°C | |
| GM2A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title