missing translation for 'onlineSavingsMsg'
Learn More

GM2A Antibody (06), DyLight 405 , Novus Biologicals™

Product Code. 18406869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406869 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406869 Supplier Novus Biologicals Supplier No. NBP306149V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GM2A Monoclonal antibody specifically detects GM2A in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM2A
Applications ELISA
Classification Monoclonal
Clone 06
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Cerebroside sulfate activator protein, ganglioside GM2 activator, GM2 ganglioside activator, GM2-AP, SAP-3GM2 ganglioside activator protein, Shingolipid activator protein 3, sphingolipid activator protein 3
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GM2A (AAA35907.1; Met1-Ile193).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2760
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.