missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2 Ganglioside Antibody (L55-81), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81278F
This item is not returnable.
View return policy
Description
GM2 Ganglioside Monoclonal specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GM2 Ganglioside | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| L55-81 binds specifically to GM2 ganglioside; this antibody shows no reactivity towards GM1, GM3, GD2 or normal tissues- the L55-81 epitope contains both a GalNAc and sialic acid bound to lactose. Ganglioside GM2 (GalNAcbeta1-4(NeuNAcalpha2-3)Galbeta1-4Glcbeta-1ceramide) is a glycosphingolipid present on the cell surface and a major tumor-differentiation antigen in human melanoma and hepatoma. | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| L55-81 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GM2 (ganglioside), Tay-Sachs ganglioside | |
| L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction