missing translation for 'onlineSavingsMsg'
Learn More

GM2 Ganglioside Antibody (L55-81), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18345660 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345660 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345660 Supplier Novus Biologicals Supplier No. NBP281278AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GM2 Ganglioside Monoclonal specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM2 Ganglioside
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone L55-81
Conjugate Alexa Fluor 532
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias GM2 (ganglioside), Tay-Sachs ganglioside
Host Species Rabbit
Immunogen L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity L55-81 binds specifically to GM2 ganglioside; this antibody shows no reactivity towards GM1, GM3, GD2 or normal tissues- the L55-81 epitope contains both a GalNAc and sialic acid bound to lactose. Ganglioside GM2 (GalNAcbeta1-4(NeuNAcalpha2-3)Galbeta1-4Glcbeta-1ceramide) is a glycosphingolipid present on the cell surface and a major tumor-differentiation antigen in human melanoma and hepatoma.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.