missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2 Ganglioside Antibody (L55-81), Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
GM2 Ganglioside Monoclonal specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | GM2 Ganglioside |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | L55-81 |
| Conjugate | Alexa Fluor 405 |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | GM2 (ganglioside), Tay-Sachs ganglioside |
| Host Species | Rabbit |
| Immunogen | L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?