Learn More
Invitrogen™ GM130 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1077
Description
PA1-077 detects GM130 from human samples. PA1-077 has been successfully used in Western blot, flow cytometry, and immunofluorescence procedures. By Western blot, PA1-077 detects a ~135 kDa band representing GM130 from Hela cell lysate. The PA1-077 immunogen is a synthetic peptide corresponding to residues G(368) Q V M E S V R Q L Q M E R D K(383) of human GM130. PA1-077 can be used with blocking peptide PEP-296.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This encoded protein has been postulated to play roles in the stacking of Golgi cisternae and in vesicular transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined.
Specifications
| GM130 | |
| Polyclonal | |
| Unconjugated | |
| Golga2 | |
| 130 kDa cis-Golgi matrix protein; AW555139; cis-Golgi matrix protein GM130; Gm130; GM130 autoantigen; GOLGA2; golgi autoantigen, golgin subfamily a, 2; Golgi matrix protein GM130; golgin A2; Golgin subfamily A member 2; Golgin subfamily A member 2; LOW QUALITY PROTEIN: golgin subfamily A member 2; golgin-95; mKIAA4150; RP11-395P17.5; SY11 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q08379 | |
| Golga2 | |
| Synthetic peptide corresponding to residues G(368) Q V M E S V R Q L Q M E R D K(383) of human GM130. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.