missing translation for 'onlineSavingsMsg'
Learn More

GM130/GOLGA2 Antibody, Novus Biologicals™

Product Code. 18380739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380739 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380739 Supplier Novus Biologicals Supplier No. H00002801B02P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GM130/GOLGA2 Polyclonal antibody specifically detects GM130/GOLGA2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM130/GOLGA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004477.2
Gene Alias 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein
Host Species Mouse
Immunogen GOLGA2 (NP_004477.2, 1 a.a. - 990 a.a.) full-length human protein. MSEETRQSKLAAAKKKLREYQQRNSPGVPTGAKKKKKIKNGSNPETTTSGGCHSPEDTPKDNAATLQPSDDTVLPGGVPSPGASLTSMAASQNHDADNVPNLMDETKTFSSTESLRQLSQQLNGLVCESATCVNGEGPASSANLKDLESRYQQLAVALDSSYVTNKQLNITIEKLKQQNQEITDQLEEEKKECHQKQGALREQLQVHIQTIGILVSEKAELQTALAHTQHAARQKEGESEDLASRLQYSRRRVGELERALSAVSTQQKKADRYNKELTKERDALRLELYKNTQSNEDLKQEKSELEEKLRVLVTEKAGMQLNLEELQKKLEMTELLLQQFSSRCEAPDANQQLQQAMEERAQLEAHLGQVMESVRQLQMERDKYAENLKGESAMWRQRMQQMSEQVHTLREEKECSMSRVQELETSLAELRNQMAEPPPPEPPAGPSEVEQQLQAEAEHLRKELEGLAGQLQAQVQDNEGLSRLNREQEERLLELERAAELWGEQAEARRQILETMQNDRTTISRALSQNRELKEQLAELQSGFVKLTNENMEITSALQSEQHVKRELGKKLGELQEKLSELKETVELKSQEAQSLQQQRDQYLGHLQQYVAAYQQLTSEKEVLHNQLLLQTQLVDQLQQQEAQGKAVAEMARQELQETQERLEAATQQNQQLRAQLSLMAHPGEGDGLDREEEEDEEEEEEEAVAVPQPMPSIPEDLESREAMVAFFNSAVASAEEEQARLRGQLKEQRVRCRRLAHLLASAQKEPEAAAPAPGTGGDSVCGETHRALQGAMEKLQSRFMELMQEKADLKERVEELEHRCIQLSGETDTIGEYIALYQSQRAVLKERHREKEEYISRLAQDKEEMKVKLLELQELVLRLVGDRNEWHGRFLAAAQNPADEPTSGAPAPQELGAANQQGDLCEVSLAGSVEPAQGEAREGSPRDNPTAQQIMQLLREMQNPRERPGLGSNPCIPFFYRADENDEVKITVI
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2801
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.